Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRTFDC1 Mouse Monoclonal Antibody [clone: 07]

Supplier: Sino Biological

14023-MM07-100 14023-MM07-50 14023-MM07-200
103654-666EA 160.46 USD InStock
103654-666 103611-438 103654-668
Anti-PRTFDC1 Mouse Monoclonal Antibody [clone: 07]
Antibodies
Anti-PRTFDC1 Antibody

Type: Primary
Antigen: PRTFDC1
Clonality: Monoclonal
Clone: Clone: 07
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty