Anti-EBAG9 Rabbit Polyclonal Antibody

Supplier: Sino Biological
14012-T60-100 14012-T60-200 14012-T60-50
103610-868EA 352.03 USD
103610-868 103588-236 103610-866
Anti-EBAG9 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human RCAS1 / EBAG9 (rh RCAS1 / EBAG9; Catalog#14012-H07E; O00559; Arg28-Ser213). RCAS1 / EBAG9 specific IgG was purified by Human RCAS1 / EBAG9 affinity chromatography.

Type: Primary
Antigen: EBAG9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR