Anti-EBAG9 Mouse Monoclonal Antibody [clone:01]

Supplier: Sino Biological
14012-MM01-50 14012-MM01-100 14012-MM01-200
103654-640EA 270.87 USD
103654-640 103654-642 103610-862
Anti-EBAG9 Mouse Monoclonal Antibody [clone:01]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RCAS1 / EBAG9 (rh RCAS1 / EBAG9; Catalog#14012-H07E; O00559; Arg28-Ser213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EBAG9
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR