Anti-ALPI Rabbit Polyclonal Antibody

Supplier: Sino Biological
13225-RP01-100 13225-RP01-400 13225-RP01-200
103650-868EA 351.49 USD
103650-868 103650-872 103650-870
Anti-ALPI Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human ALPI (rh ALPI; Catalog#13225-H08H; P09923; Met1-Asp503). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR