Anti-PHGDH Mouse Monoclonal Antibody [clone:05]

Supplier: Sino Biological
13167-MM05-200 13167-MM05-100 13167-MM05-50
103610-188EA 142.96 USD
103610-188 103650-644 103650-642
Anti-PHGDH Mouse Monoclonal Antibody [clone:05]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PGDH / PHGDH (rh PGDH / PHGDH; Catalog#13167-H08E; O43175; Met1-Phe533). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.

Type: Primary
Antigen: PHGDH
Clonality: Monoclonal
Clone: 05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR