Anti-SCGN Mouse Monoclonal Antibody [clone:02]

Supplier: Sino Biological
12411-MM02-200 12411-MM02-50 12411-MM02-100 12411-MM02-1
103609-454EA 142.96 USD
103609-454 103636-930 103637-512 103587-678
Anti-SCGN Mouse Monoclonal Antibody [clone:02]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SCGN / Secretagogin (rh SCGN / Secretagogin; Catalog#12411-H07E; NP_008929.2; Asp2-Pro276). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SCGN
Clonality: Monoclonal
Clone: 02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR