Anti-SPINT1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11742-RP02-50 11742-RP02-100 11742-RP02-200
103608-622EA 142.96 USD
103608-622 103635-582 103635-580
Anti-SPINT1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human HAI-1 / SPINT1 (rh HAI-1 / SPINT1; Catalog#11742-H08H; O43278-2; Met 1-Val 433). HAI-1 / SPINT1 specific IgG was purified by Human HAI-1 / SPINT1 affinity chromatography.

Type: Primary
Antigen: HAI-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR