Anti-STIM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11434-RP02-50 11434-RP02-100 11434-RP02-200
103645-038EA 142.96 USD
103645-038 103634-186 103634-188
Anti-STIM1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human STIM1 (rh STIM1; Catalog#11434-H08H; NP_003147.2; Met 1-Asp 213). STIM1 specific IgG was purified by Human STIM1 affinity chromatography
.

Type: Primary
Antigen: STIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR