Anti-PMP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11426-RP01-400 11426-RP01-100 11426-RP01-200
103635-294EA 562.97 USD
103635-294 103635-292 103635-290
Anti-PMP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PMP2 (rh PMP2; Catalog#11426-H07E; NP_002668.1; Ser 2-Val 132). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: PMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR