Anti-HYOU1 Rabbit Monoclonal Antibody [clone: 201]

Supplier: Sino Biological
11342-R201-50 11342-R201-100
103635-264EA 271.4 USD
103635-264 103644-970
Anti-HYOU1 Rabbit Monoclonal Antibody [clone: 201]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human ORP150 / HYOU1 / HSP12A (rh ORP150 / HYOU1 / HSP12A; Catalog#11342-H08H; NP_001124463.1; Met 695-Leu 994).

Type: Primary
Antigen: ORP150
Clonality: Monoclonal
Clone: 201
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR