Anti-FUT10 Rabbit Monoclonal Antibody [clone: 003]

Supplier: Sino Biological
11327-R003-50 11327-R003-100
103644-960EA 150.18 USD
103644-960 103633-828
Anti-FUT10 Rabbit Monoclonal Antibody [clone: 003]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human FUT10 (rh FUT10; Catalog#11327-H07H; Q6P4F1-1; Leu 32-Asp 479).

Type: Primary
Antigen: FUT10
Clonality: Monoclonal
Clone: 003
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR