Anti-SAA4 Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
11311-R001-100 11311-R001-50
103643-658EA 150.18 USD
103643-658 103633-708
Anti-SAA4 Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human SAA4 (rh SAA4; Catalog#11311-H09E; P35542; Glu19-Tyr130).

Type: Primary
Antigen: SAA4
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR