Anti-CLU Rabbit Polyclonal Antibody

Supplier: Sino Biological
11297-T04-50 11297-T04-200 11297-T04-100
103644-928EA 352.03 USD
103644-928 103587-190 103644-926
Anti-CLU Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human Clusterin/Apolipoprotein J/Apo-J (rh Clusterin/Apolipoprotein J/Apo-J; Catalog#11297-H08H; NP_001822.2; Met1-Glu501). Clusterin/Apolipoprotein J/Apo-J specific IgG was purified by Human Clusterin/Apolipoprotein J/Apo-J affinity chromatography.

Type: Primary
Antigen: Clusterin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR