Anti-CD96 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11202-RP02-200 11202-RP02-50 11202-RP02-100
103644-648EA 142.96 USD
103644-648 103635-114 103635-112
Anti-CD96 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CD96 (rh CD96; Catalog#11202-H08H; P40200-2; Met 1-Met 503). CD96 specific IgG was purified by Human CD96 affinity chromatography.

Type: Primary
Antigen: CD96
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR