Anti-CD96 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11202-RP01-200 11202-RP01-400 11202-RP01-100
103635-110EA 562.97 USD
103635-110 103635-108 103635-106
Anti-CD96 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CD96 (rh CD96; Catalog#11202-H08H; P40200-2; Met1-Met503). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD96
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR