Anti-S100A12 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11143-RP02-50 11143-RP02-100 11143-RP02-200
103644-514EA 142.96 USD
103644-514 103632-870 103632-872
Anti-S100A12 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human S100A12 (rh S100A12; Catalog#11143-HNAE; NP_005612.1; Met 1-Glu 92). S100A12 specific IgG was purified by Human S100A12 affinity chromatography
.

Type: Primary
Antigen: S100A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR