Anti-S100A12 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11143-RP01-100 11143-RP01-400 11143-RP01-200
103632-866EA 351.49 USD
103632-866 103632-868 103632-864
Anti-S100A12 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human S100A12 (rh S100A12; Catalog#11143-HNAE; NP_005612.1; Met 1-Glu 92). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: S100A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR