Anti-NGF Rabbit Monoclonal Antibody [clone:B64]

Supplier: Sino Biological
11050-RB64-50 11050-RB64-100 11050-RB64-1
103619-154EA 301.66 USD
103619-154 103587-006 103644-130
Anti-NGF Rabbit Monoclonal Antibody [clone:B64]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human NGF / NGFB / beta-NGF (rh NGF / NGFB / beta-NGF; Catalog#11050-HNAC; NP_002497.2; Ser122-Arg239).

Type: Primary
Antigen: NGF
Clonality: Monoclonal
Clone: B64
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR