Anti-HA Rabbit Polyclonal Antibody

Supplier: Sino Biological
11048-RP02-50
103644-108EA 162.95 USD
103644-108
Anti-HA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Influenza A virus H5N1 (Avian Flu) Hemagglutinin extracellular domain. Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Hemagglutinin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: H5N1 (A/Anhui/1/2005) HA
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR