Anti-HA Rabbit Monoclonal Antibody [clone: 89]

Supplier: Sino Biological
11048-RM09-50 11048-RM09-100
103644-106EA 203.66 USD
103644-106 103619-126
Anti-HA Rabbit Monoclonal Antibody [clone: 89]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A virus H5N1 (Avian Flu) Hemagglutinin extracellular domain.

Type: Primary
Antigen: Hemagglutinin
Clonality: Monoclonal
Clone: 89
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: H5N1 (A/Anhui/1/2005) HA
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR