Anti-HA Rabbit Monoclonal Antibody [clone: 82]

Supplier: Sino Biological
11048-RM07-100 11048-RM07-50 11048-RM07-20
103644-102EA 203.66 USD
103644-102 76777-392 103619-122
Anti-HA Rabbit Monoclonal Antibody [clone: 82]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A virus H5N1 (Avian Flu) Hemagglutinin extracellular domain.

Type: Primary
Antigen: Hemagglutinin
Clonality: Monoclonal
Clone: 82
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: H5N1 (A/Anhui/1/2005) HA H5N1 (A/Anhui/1/2005) HA2 Sub-unit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR