Anti-FCGR3 Mouse Monoclonal Antibody [clone: 10] (FITC (Fluorescein Isothiocyanate))

Supplier: Sino Biological
11046-MM10-F-100 11046-MM10-F-25
103631-620EA 151.5 USD
103631-620 103629-404
Anti-FCGR3 Mouse Monoclonal Antibody [clone: 10] (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD16b/Fc gamma RIIIb (rh CD16b/Fc gamma RIIIb; Catalog#11046-H08H; O75015.2; Met1-Ser200) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: Clone: 10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR