Anti-FCGR3B Mouse Monoclonal Antibody [clone:2D2G5B9]

Supplier: Sino Biological
11046-MM01-50 11046-MM01-100 11046-MM01-200
103619-018EA 352.03 USD
103619-018 103619-016 103644-076
Anti-FCGR3B Mouse Monoclonal Antibody [clone:2D2G5B9]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD16b (rh CD16b; Catalog#11046-H08H; O75015.2; Met 1-Ser 200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD16b
Clonality: Monoclonal
Clone: 2D2G5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR