Anti-PARP1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11040-RP01-400 11040-RP01-100 11040-RP01-200
103619-004EA 562.97 USD
103619-004 103620-396 103620-394
Anti-PARP1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PARP1 (rh PARP1; Catalog#11040-H08B; NP_001609.2; Met 1-Trp 1014). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PARP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR