Anti-Insulin Rabbit Polyclonal Antibody

11038-T08-50 11038-T08-200 11038-T08-100
103586-990EA 106.62 USD
103586-990 103644-054 103644-052
Anti-Insulin Rabbit Polyclonal Antibody
Anti-Insulin Antibody

Type: Primary
Antigen: Insulin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR