Anti-CD9 Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological
11029-MM01-50 11029-MM01-100 11029-MM01-200
103618-974EA 352.03 USD
103618-974 103618-972 103644-040
Anti-CD9 Mouse Monoclonal Antibody [clone: 01]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD9 (rh CD9; Catalog#11029-H08H; NP_001760.1; Ser 112-Ile 195). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR