Anti-CD5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11027-RP02-100 11027-RP02-50 11027-RP02-200
103618-204EA 352.03 USD
103618-204 103644-028 103618-202
Anti-CD5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CD5 (rh CD5; Catalog#11027-H08H; NP_055022.2; Met 1-Pro 372). CD5 specific IgG was purified by Human CD5 affinity chromatography.

Type: Primary
Antigen: CD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR