Anti-CD5 Rabbit Monoclonal Antibody [clone: 322]

Supplier: Sino Biological
11027-R322-100 11027-R322-50
103644-026EA 150.18 USD
103644-026 103618-962
Anti-CD5 Rabbit Monoclonal Antibody [clone: 322]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD5 / Cluster of Differentiation 5 (rh CD5 / Cluster of Differentiation 5; Catalog#11027-H08H; NP_055022.2; Met1-Pro372).

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 322
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR