Anti-CD5 Rabbit Monoclonal Antibody [clone: 028] (PE (Phycoerythrin))

Supplier: Sino Biological
11027-R028-P-100 11027-R028-P-25
103629-374EA 70.7 USD
103629-374 103631-486
Anti-CD5 Rabbit Monoclonal Antibody [clone: 028] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD5 (rh CD5; Catalog#11027-H08H; NP_055022.2; Met1-Pro372) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: Clone: 028
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR