Anti-IL22RA2 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological
11025-R001-50 11025-R001-1 11025-R001-100
103586-982EA 1500.59 USD
103586-982 103644-014 103618-940
Anti-IL22RA2 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-22BP/IL-22RA2 (rh IL-22BP/IL-22RA2; Catalog#11025-H08H; NP_851826.1; Met1-Pro231).

Type: Primary
Antigen: IL-22BP
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR