Anti-TF Rabbit Polyclonal Antibody

Supplier: Sino Biological
11019-RP01-400 11019-RP01-100 11019-RP01-200
103618-902EA 351.49 USD
103618-902 103618-900 103618-904
Anti-TF Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human Transferrin / TF (rh Transferrin; Catalog#11019-H08H; NP_001054.1; Met 1-Pro 698). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: Transferrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR