Anti-TF Mouse Monoclonal Antibody [clone:6G5C11G2]

Supplier: Sino Biological
11019-MM03-50 11019-MM03-100 11019-MM03-200
103618-892EA 213.93 USD
103618-892 103643-990 103618-890
Anti-TF Mouse Monoclonal Antibody [clone:6G5C11G2]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Transferrin / TF (rh Transferrin / TF; Catalog#11019-H08H; NP_001054.1; Met 1-Pro 698). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Transferrin
Clonality: Monoclonal
Clone: 6G5C11G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR