Anti-SerpinI2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11008-RP02-100 11008-RP02-200 11008-RP02-50
103618-852EA 352.03 USD
103618-852 103618-850 103643-980
Anti-SerpinI2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human SerpinI2 (rh SerpinI2; Catalog#11008-H08H; NP_006208.1; Met 1-Leu 405). SerpinI2 specific IgG was purified by Human SerpinI2 affinity chromatography.

Type: Primary
Antigen: SERPINI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR