Anti-SerpinI2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11008-RP01-200 11008-RP01-100 11008-RP01-400
103618-846EA 351.49 USD
103618-846 103618-844 103618-848
Anti-SerpinI2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human SerpinI2 (rh SerpinI2; Catalog#11008-H08H; NP_006208.1; Met 1-Leu 405). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SERPINI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR