Anti-MS4A1 Rabbit Monoclonal Antibody [clone: 001] (PE (Phycoerythrin))

Supplier: Sino Biological
11007-R001-P-100 11007-R001-P-25
103629-348EA 70.7 USD
103629-348 103631-452
Anti-MS4A1 Rabbit Monoclonal Antibody [clone: 001] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, Human CD20 Transfected CHO Cells and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR