Anti-SerpinA6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
10998-RP01-100 10998-RP01-400 10998-RP01-200
103618-814EA 270.2 USD
103618-814 103618-812 103618-816
Anti-SerpinA6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human SerpinA6 (rh SERPINA6; Catalog#10998-H08H; NP_001747.2; Met1-Val405). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CBG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR