Anti-SerpinG1 Mouse Monoclonal Antibody [clone:06]

Supplier: Sino Biological
10995-MM06-200 10995-MM06-100 10995-MM06-50 10995-MM06-1
103643-946EA 142.96 USD
103643-946 103586-962 103620-384 103620-382
Anti-SerpinG1 Mouse Monoclonal Antibody [clone:06]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human C1 inhibitor / SerpinG1 (rh C1 inhibitor / SerpinG1; Catalog#10995-H08H; NP_000053.2; Met1-Ala500). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SERPING1
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR