Anti-SERPINA7 Mouse Monoclonal Antibody [clone:02]

Supplier: Sino Biological
10993-MM02-200 10993-MM02-100 10993-MM02-50
103618-766EA 352.03 USD
103618-766 103618-764 103643-928
Anti-SERPINA7 Mouse Monoclonal Antibody [clone:02]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinA7 (rh SerpinA7; Catalog#10993-H08H; NP_000345.2; Met1-Ala415). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SERPINA7
Clonality: Monoclonal
Clone: 02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR