Anti-S100A13 Rabbit Polyclonal Antibody

Supplier: Sino Biological
10991-RP02-100 10991-RP02-50 10991-RP02-200
103618-762EA 352.03 USD
103618-762 103618-760 103643-926
Anti-S100A13 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human S100A13 (rh S100A13; Catalog#10991-HNAE; NP_001019381.1; Met 1-Lys 98). S100A13 specific IgG was purified by Human S100A13 affinity chromatography
.

Type: Primary
Antigen: S100A13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR