Anti-CD2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
10982-RP02-50 10982-RP02-100 10982-RP02-200
103618-736EA 352.03 USD
103618-736 103618-734 103643-642
Anti-CD2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CD2 (rh CD2; Catalog#10982-H08H; NP_001758.2; Met 1-Asp 209). CD2 specific IgG was purified by Human CD2 affinity chromatography.

Type: Primary
Antigen: CD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR