Anti-CD8A Mouse Monoclonal Antibody [clone: 2C5D4C8]

Supplier: Sino Biological
10980-MM04-200 10980-MM04-50 10980-MM04-100
103618-666EA 270.87 USD
103618-666 103643-872 103620-372
Anti-CD8A Mouse Monoclonal Antibody [clone: 2C5D4C8]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD8A (rh CD8A; Catalog#10980-H08H; NP_001759.3; Met 1-Asp 182). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 2C5D4C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR