Anti-CD3E Rabbit Polyclonal Antibody

Supplier: Sino Biological
10977-T20-50 10977-T20-200 10977-T20-100
103643-868EA 352.03 USD
103643-868 103643-866 103586-944
Anti-CD3E Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CD3e / CD3 epsilon (rh CD3e / CD3 epsilon; Catalog#10977-H08H; NP_000724.1; Met1-Asp126). CD3e / CD3 epsilon specific IgG was purified by Human CD3e / CD3 epsilon affinity chromatography.

Type: Primary
Antigen: CD3e
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR