Anti-IL7R Mouse Monoclonal Antibody [clone:5H10B5D6]

Supplier: Sino Biological
10975-MM12-200 10975-MM12-50 10975-MM12-100
103643-840EA 142.96 USD
103643-840 103618-630 103618-628
Anti-IL7R Mouse Monoclonal Antibody [clone:5H10B5D6]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL7RA (rh IL7RA; Catalog#10975-H08H; AAC83204.1; Met 1-Gly 236). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: 5H10B5D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR