Anti-SDF2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
10950-T16-50 10950-T16-100 10950-T16-200
103643-634EA 352.03 USD
103643-634 103643-632 103586-924
Anti-SDF2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human SDF-2/SDF2 (rh SDF-2/SDF2; Catalog#10950-H08B; NP_008854.2; Met1-Leu211). SDF-2/SDF2 specific IgG was purified by Human SDF-2/SDF2 affinity chromatography.

Type: Primary
Antigen: SDF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR