Anti-IL10 Mouse Monoclonal Antibody [clone: 03]

Supplier: Sino Biological
10947-MM03-200 10947-MM03-50 10947-MM03-100 10947-MM03-1
103643-800EA 142.96 USD
103643-800 103618-532 103618-530 103586-920
Anti-IL10 Mouse Monoclonal Antibody [clone: 03]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL10/IL-10/Interleukin-10 (rh IL10/IL-10/Interleukin-10; Catalog#10947-HNAE; NP_000563.1; Ser 19-Asn 178). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: 03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR