Anti-IL16 Rabbit Monoclonal Antibody [clone: 373]

Supplier: Sino Biological
10944-R373-100 10944-R373-50
103643-776EA 150.18 USD
103643-776 103618-498
Anti-IL16 Rabbit Monoclonal Antibody [clone: 373]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-16 (rh IL-16; Catalog#10944-H07E; AAC12732.1; Pro 2-Ser 130).

Type: Primary
Antigen: IL-16
Clonality: Monoclonal
Clone: 373
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR