Anti-IL13RA1 Rabbit Polyclonal Antibody

10943-T16-200 10943-T16-100 10943-T16-50
103643-768EA 214.31 USD
103643-768 103643-766 103586-906
Anti-IL13RA1 Rabbit Polyclonal Antibody
Anti-IL13RA1 Antibody

Type: Primary
Antigen: IL13RA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR