Anti-PBK Rabbit Polyclonal Antibody

Supplier: Sino Biological
10729-RP01-200 10729-RP01-400 10729-RP01-100
103620-090EA 351.49 USD
103620-090 103620-092 103620-088
Anti-PBK Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human PBK / TOPK (rh PBK; Catalog#10729-H08B; NP_060962.2; Met 1-Val 322). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PBK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR