Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: 2F8B1]

Supplier: Sino Biological
10413-MM04-100 10413-MM04-50 10413-MM04-200
103649-172EA 270.87 USD
103649-172 103641-778 103649-174
Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: 2F8B1]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TRAIL R4 / CD264 / TNFRSF10D (rh TRAIL R4 / CD264 / TNFRSF10D; Catalog#10413-H08H; NP_003831.2; Met 1-His 211). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: DcR2
Clonality: Monoclonal
Clone: 2F8B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR