Anti-EphB6 Rabbit Monoclonal Antibody [clone: 037]

Supplier: Sino Biological
10197-R037-100 10197-R037-50
103646-474EA 271.4 USD
103646-474 103640-412
Anti-EphB6 Rabbit Monoclonal Antibody [clone: 037]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB6 (rh EphB6; Catalog#10197-H08H; NP_004436.1; Met 1-Ser 579).

Type: Primary
Antigen: EphB6
Clonality: Monoclonal
Clone: 037
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR